siRNA Details
| Gene Symbol: Zfyve26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008550.1 | NP_001008550.1 | 33, 34 | 6284 | ||
| GenBank mRNA | AK029679.1 | - | 1, 2 | 47 | ||
| AK042386.1 | - | 7, 8 | 1627 | |||
| AK169541.1 | - | 7, 8 | 1625 | |||
| AK220333.1 | - | 33, 34 | 6255 | |||
| BC141359.1 | - | 33, 34 | 6267 | 9055971 | ||

Zfyve26
Genomic Map