siRNA Details
| Gene Symbol: Syne3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042699.2 | NP_001036164.1 | 15 | 2617 | ||
| NM_172500.3 | NP_766088.2 | 13 | 2262 | |||
| GenBank mRNA | AK029216.1 | - | 13 | 2264 | ||
| AK030542.1 | - | 2 | 255 | |||
| AK166152.1 | - | 2 | 129 | |||
| BC099694.1 | - | 15 | 2617 | 4507746 | ||

Syne3
Genomic Map