siRNA Details
Gene Symbol: Zswim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172503.3 | NP_766091.2 | 14 | 3261 | ||
GenBank mRNA | AK052168.1 | - | 14 | 3275 | ||
AK053663.1 | - | 14 | 3275 | |||
AK131119.1 | - | 9 | 2119 | |||
BC016518.1 | - | 4 | 1078 | |||
BC058080.1 | - | 14 | 3261 | 6822376 |
