siRNA Details
| Gene Symbol: Mtg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199301.2 | NP_955005.2 | 9 | 808 | ||
| GenBank mRNA | AK144453.1 | - | 9 | 733 | ||
| AK144625.1 | - | 9 | 766 | |||
| BC027306.1 | - | 9 | 717 | 4019717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtg1| Gene Symbol: Mtg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199301.2 | NP_955005.2 | 9 | 808 | ||
| GenBank mRNA | AK144453.1 | - | 9 | 733 | ||
| AK144625.1 | - | 9 | 766 | |||
| BC027306.1 | - | 9 | 717 | 4019717 | ||