siRNA Details
| Gene Symbol: Rho | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145383.1 | NP_663358.1 | 4 | 893 | ||
| GenBank mRNA | AK044333.1 | - | 4 | 913 | ||
| AK044412.1 | - | 4 | 911 | |||
| BC013125.1 | - | 4 | 893 | 4500760 | ||
| BC031766.1 | - | 4 | 894 | 4527040 | ||
More Information
Set Membership: |
|

Rho
Genomic Map