siRNA Details
| Gene Symbol: BC027231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145972.4 | NP_666084.1 | 5, 6 | 611 | ||
| GenBank mRNA | AK137274.1 | - | 4, 5 | 1304 | ||
| AK144612.1 | - | 5, 6 | 592 | |||
| AK162291.1 | - | 4, 5 | 453 | |||
| BC027231.1 | - | 5, 6 | 614 | 3585175 | ||

BC027231
Genomic Map