siRNA Details
| Gene Symbol: Kctd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172509.3 | NP_766097.1 | 4 | 700 | ||
| GenBank mRNA | AK029942.1 | - | 4 | 675 | ||
| AK034583.1 | - | 4 | 676 | |||
| AK083583.1 | - | 4 | 701 | |||
| AK140606.1 | - | 4 | 675 | |||
| AK162568.1 | - | 4 | 729 | |||
| BC051544.1 | - | 3 | 243 | |||
| BC139070.1 | - | 4 | 602 | 8862092 | ||
| BC141201.1 | - | 4 | 602 | 9055813 | ||

Kctd7
Genomic Map