siRNA Details
| Gene Symbol: Wbscr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145218.3 | NP_660253.2 | 11 | 2704 | ||
| GenBank mRNA | AB175684.1 | - | 11 | 1782 | ||
| AF467979.1 | - | 11 | 1985 | |||
| BC052469.1 | - | 11 | 2100 | 6810287 | ||
| BC069177.1 | - | 11 | 2704 | |||
| BC158110.1 | - | 11 | 2731 | 9088184 | ||

Wbscr17
Genomic Map