siRNA Details
Gene Symbol: Abhd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272070.1 | NP_001258999.1 | 3, 4 | 482 | ||
NM_172511.4 | NP_766099.3 | 3, 4 | 482 | |||
GenBank mRNA | AK052670.1 | - | 3, 4 | 431 | ||
AK084998.1 | - | 3, 4 | 430 | |||
BC058347.1 | - | 3, 4 | 431 | 6820515 | ||
CX237340.1 | - | 2, 3 | 245 |
