siRNA Details
| Gene Symbol: Abhd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272070.1 | NP_001258999.1 | 2, 3 | 360 | ||
| NM_172511.4 | NP_766099.3 | 2, 3 | 360 | |||
| GenBank mRNA | AK052670.1 | - | 2, 3 | 309 | ||
| AK084998.1 | - | 2, 3 | 308 | |||
| BC058347.1 | - | 2, 3 | 309 | 6820515 | ||
| BY173258.1 | - | 2, 3 | 312 | |||
| BY281009.1 | - | 2, 3 | 361 | |||

Abhd10
Genomic Map