siRNA Details
Gene Symbol: Evi5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039578.3 | NP_001034667.3 | 2 | 347 | ||
GenBank mRNA | AK045240.1 | - | 2 | 348 | ||
AK087482.1 | - | 2 | 348 | |||
AK193949.1 | - | 2 | 251 | |||
BC042653.1 | - | 2 | 327 |
