siRNA Details
Gene Symbol: Fam126b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172513.3 | NP_766101.3 | 12 | 1606 | ||
GenBank mRNA | AK052638.1 | - | 12 | 1533 | ||
AK144602.1 | - | 13 | 1711 | |||
AK168372.1 | - | 12 | 1596 | |||
BC049806.1 | - | 12 | 1542 | |||
BC068302.1 | - | 12 | 1514 | 30431671 |
