siRNA Details
| Gene Symbol: Fstl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253719.1 | NP_001240648.1 | 7 | 927 | ||
| NM_178673.4 | NP_848788.2 | 7 | 1187 | |||
| GenBank mRNA | AF374460.1 | - | 7 | 1161 | ||
| AK033652.1 | - | 7 | 1178 | |||
| AK046070.1 | - | 7 | 928 | |||
| AK081152.1 | - | 7 | 1178 | |||
| AK138388.1 | - | 7 | 1479 | |||
| AK140611.1 | - | 7 | 1190 | |||
| BC075640.1 | - | 7 | 1150 | 30545529 | ||

Fstl5
Genomic Map