siRNA Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198021.2 | NP_932138.1 | 17 | 2404 | ||
| GenBank mRNA | AK136092.1 | - | 2 | 140 | ||
| AK139747.1 | - | 9 | 990 | |||
| AK139761.1 | - | 17 | 2388 | |||
| BC042443.1 | - | 17 | 2404 | 4037878 | ||

Scyl2
Genomic Map