siRNA Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198021.2 | NP_932138.1 | 2 | 410 | ||
| GenBank mRNA | AK052108.1 | - | 2 | 392 | ||
| AK139761.1 | - | 2 | 394 | |||
| AK167276.1 | - | 2 | 394 | |||
| BC042443.1 | - | 2 | 410 | 4037878 | ||

Scyl2
Genomic Map