siRNA Details
| Gene Symbol: Gm732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033252.2 | NP_001028424.1 | 2 | 1351 | ||
| GenBank mRNA | AK132985.1 | - | 2 | 1353 | ||
| BC147603.1 | - | 2 | 1290 | 9056593 | ||
| BC147609.1 | - | 2 | 1290 | 9056599 | ||

Gm732
Genomic Map