siRNA Details
| Gene Symbol: Dis3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001295.2 | NP_001001295.2 | 4 | 640 | ||
| NM_001177784.1 | NP_001171255.1 | 4 | 601 | |||
| NM_172519.3 | NP_766107.1 | 4 | 544 | |||
| GenBank mRNA | AK029974.1 | - | 4 | 545 | ||
| AK032451.1 | - | 4 | 549 | |||
| AK035357.1 | - | 4 | 540 | |||
| AK077031.1 | - | 4 | 604 | |||
| AK150851.1 | - | 4 | 590 | |||
| AK151780.1 | - | 4 | 590 | |||
| AK153433.1 | - | 4 | 590 | |||
| AK173310.1 | - | 4 | 574 | |||
| BC056939.1 | - | 4 | 573 | 5708685 | ||

Dis3l
Genomic Map