siRNA Details
| Gene Symbol: Dis3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001295.2 | NP_001001295.2 | 12 | 2016 | ||
| NM_001177784.1 | NP_001171255.1 | 12 | 1977 | |||
| NM_172519.3 | NP_766107.1 | 12 | 1920 | |||
| GenBank mRNA | AK029974.1 | - | 12 | 1921 | ||
| AK032451.1 | - | 12 | 1925 | |||
| AK077031.1 | - | 12 | 1980 | |||
| AK150851.1 | - | 12 | 1962 | |||
| AK151780.1 | - | 12 | 1962 | |||
| AK153433.1 | - | 12 | 1962 | |||
| AK173310.1 | - | 12 | 1950 | |||
| BC056939.1 | - | 12 | 1949 | 5708685 | ||

Dis3l
Genomic Map