siRNA Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009320.4 | NP_033346.2 | 8 | 1229 | ||
| GenBank mRNA | AK143510.1 | - | 8 | 1193 | ||
| BC015245.1 | - | 8 | 1166 | 4242193 | ||
| L03292.2 | - | 8 | 1201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a6| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009320.4 | NP_033346.2 | 8 | 1229 | ||
| GenBank mRNA | AK143510.1 | - | 8 | 1193 | ||
| BC015245.1 | - | 8 | 1166 | 4242193 | ||
| L03292.2 | - | 8 | 1201 | |||