siRNA Details
| Gene Symbol: Tbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013684.3 | NP_038712.3 | 4, 5 | 768 | ||
| GenBank mRNA | AK085037.1 | - | 4, 5 | 769 | ||
| BC012685.1 | - | 6, 7 | 1004 | 3498542 | ||
| BC016476.1 | - | 4, 5 | 746 | 3584341 | ||
| BC050136.1 | - | 4, 5 | 719 | 30020603 | ||
| D01034.1 | - | 3, 4 | 585 | |||
| U63933.1 | - | 5, 6 | 774 | |||

Tbp
Genomic Map