siRNA Details
| Gene Symbol: Zfp598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183149.1 | NP_898972.1 | 4 | 535 | ||
| GenBank mRNA | AK131127.1 | - | 4 | 554 | ||
| AK138860.1 | - | 2 | 114 | |||
| AK209503.1 | - | 2 | 213 | |||
| AK220149.1 | - | 3 | 228 | |||
| BC024690.1 | - | 4 | 329 | |||
| BC050859.1 | - | 4 | 535 | 6328878 | ||

Zfp598
Genomic Map