siRNA Details
| Gene Symbol: Tbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009322.3 | NP_033348.2 | 2 | 1081 | ||
| GenBank mRNA | BC052737.1 | - | 2 | 1081 | 6825394 | |
| BC058399.1 | - | 2 | 985 | 6826831 | ||
| U49251.1 | - | 2 | 921 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbr1| Gene Symbol: Tbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009322.3 | NP_033348.2 | 2 | 1081 | ||
| GenBank mRNA | BC052737.1 | - | 2 | 1081 | 6825394 | |
| BC058399.1 | - | 2 | 985 | 6826831 | ||
| U49251.1 | - | 2 | 921 | |||