siRNA Details
| Gene Symbol: Tbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011536.2 | NP_035666.2 | 8 | 1361 | ||
| NM_172798.1 | NP_766386.1 | 7 | 1311 | |||
| GenBank mRNA | AB221665.1 | - | 8 | 1361 | ||
| AK028407.1 | - | 7 | 1245 | |||
| AK139949.1 | - | 7 | 1285 | |||
| AK147984.1 | - | 7 | 1229 | |||
| AY075134.1 | - | 8 | 1361 | |||

Tbx4
Genomic Map