siRNA Details
| Gene Symbol: Tbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011537.3 | NP_035667.1 | 8 | 1498 | ||
| GenBank mRNA | AF140427.1 | - | 8 | 1263 | ||
| AK044843.1 | - | 4 | 477 | |||
| BC090639.1 | - | 8 | 1502 | 30548234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx5| Gene Symbol: Tbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011537.3 | NP_035667.1 | 8 | 1498 | ||
| GenBank mRNA | AF140427.1 | - | 8 | 1263 | ||
| AK044843.1 | - | 4 | 477 | |||
| BC090639.1 | - | 8 | 1502 | 30548234 | ||