siRNA Details
Gene Symbol: Tbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011537.3 | NP_035667.1 | 7, 8 | 1397 | ||
GenBank mRNA | AF140427.1 | - | 7, 8 | 1162 | ||
AK044843.1 | - | 3, 4 | 376 | |||
BC090639.1 | - | 7, 8 | 1401 | 30548234 |
