siRNA Details
| Gene Symbol: Tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011539.3 | NP_035669.3 | 8 | 908 | ||
| GenBank mRNA | AK048544.1 | - | 8 | 890 | ||
| AK085414.1 | - | 5 | 659 | |||
| AK139479.1 | - | 8 | 871 | |||
| BC029014.1 | - | 8 | 910 | 5007629 | ||
| L18868.1 | - | 8 | 779 | |||

Tbxas1
Genomic Map