siRNA Details
| Gene Symbol: Fbxw10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033669.2 | NP_001028841.2 | 13 | 2737 | ||
| NM_001291441.1 | NP_001278370.1 | 13 | 2752 | |||
| GenBank mRNA | AK132680.1 | - | 13 | 2701 | ||
| BC141212.1 | - | 13 | 2743 | 9055824 | ||
| BY360237.1 | - | 1 | 47 | |||

Fbxw10
Genomic Map