siRNA Details
Gene Symbol: Ccdc186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170757.1 | NP_739563.1 | 15 | 2699 | ||
GenBank mRNA | AF349751.1 | - | 15 | 2699 | ||
AK048386.1 | - | 1 | 567 | |||
BC046394.1 | - | 3 | 303 |
