siRNA Details
| Gene Symbol: Dnajc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172338.2 | NP_758841.1 | 13, 14 | 1847 | ||
| GenBank mRNA | AK028692.1 | - | 13, 14 | 1852 | ||
| AK051048.1 | - | 13, 14 | 1848 | |||
| AK122407.1 | - | 2094 | ||||
| AK154946.1 | - | 1868 | ||||
| AK164023.1 | - | 13, 14 | 1863 | |||
| BC046424.1 | - | 7, 8 | 842 | 3989412 | ||

Dnajc16
Genomic Map