siRNA Details
| Gene Symbol: Nipal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172524.3 | NP_766112.1 | 2 | 305 | ||
| GenBank mRNA | AK035561.1 | - | 2 | 306 | ||
| AK035736.1 | - | 2 | 282 | |||
| BC139819.1 | - | 2 | 296 | 40141293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nipal4| Gene Symbol: Nipal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172524.3 | NP_766112.1 | 2 | 305 | ||
| GenBank mRNA | AK035561.1 | - | 2 | 306 | ||
| AK035736.1 | - | 2 | 282 | |||
| BC139819.1 | - | 2 | 296 | 40141293 | ||