siRNA Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083967.1 | NP_001077436.1 | 7 | 676 | ||
| NM_013685.2 | NP_038713.1 | 7 | 991 | |||
| GenBank mRNA | AK028748.1 | - | 7 | 602 | ||
| AK051958.1 | - | 6 | 899 | |||
| AK081012.1 | - | 7 | 628 | |||
| AK133885.1 | - | 8 | 1403 | |||
| BC043050.1 | - | 6 | 451 | 5708370 | ||
| S75870.1 | - | 7 | 563 | |||
| U16322.1 | - | 7 | 963 | |||

Tcf4
Genomic Map