siRNA Details
| Gene Symbol: Ago3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153402.2 | NP_700451.2 | 4 | 655 | ||
| GenBank mRNA | AB081473.1 | - | 4 | 507 | ||
| AK035042.1 | - | 4 | 621 | |||
| AK042049.1 | - | 4 | 577 | |||
| AK083652.1 | - | 4 | 622 | |||
| AK154436.1 | - | 4 | 656 | |||
| AK171118.1 | - | 4 | 656 | |||
| AK172226.1 | - | 4 | 577 | |||
| BC137964.1 | - | 4 | 502 | 8860983 | ||
| BC145061.1 | - | 4 | 502 | |||
| BC145063.1 | - | 4 | 502 | |||

Ago3
Genomic Map