siRNA Details
| Gene Symbol: Kmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081049.1 | NP_001074518.1 | 3 | 1075 | ||
| GenBank mRNA | AK145484.1 | - | 3 | 665 | ||
| EF410110.1 | - | 903 | ||||
| L17069.1 | - | 3 | 777 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kmt2a| Gene Symbol: Kmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081049.1 | NP_001074518.1 | 3 | 1075 | ||
| GenBank mRNA | AK145484.1 | - | 3 | 665 | ||
| EF410110.1 | - | 903 | ||||
| L17069.1 | - | 3 | 777 | |||