siRNA Details
| Gene Symbol: Kmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081049.1 | NP_001074518.1 | 5 | 3530 | ||
| GenBank mRNA | AK140434.1 | - | 3 | 2274 | ||
| AK140439.1 | - | 3 | 2273 | |||
| AK145484.1 | - | 5 | 3121 | |||
| AK149341.1 | - | 3 | 2417 | |||
| AK220457.1 | - | 3 | 1164 | |||
| L17069.1 | - | 5 | 3232 | |||

Kmt2a
Genomic Map