siRNA Details
| Gene Symbol: Tfap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025305.2 | NP_001020476.1 | 3 | 853 | ||
| NM_001286340.1 | NP_001273269.1 | 3 | 818 | |||
| NM_009334.3 | NP_033360.2 | 3 | 818 | |||
| GenBank mRNA | AK005128.1 | - | 3 | 776 | ||
| AK017373.1 | - | 3 | 820 | |||
| AK028906.1 | - | 3 | 773 | |||
| AK032045.1 | - | 3 | 855 | |||
| AK078046.1 | - | 2 | 508 | |||
| X78197.1 | - | 3 | 744 | |||

Tfap2b
Genomic Map