siRNA Details
| Gene Symbol: Disp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170593.3 | NP_733481.1 | 8 | 1575 | ||
| GenBank mRNA | AK122547.1 | - | 1 | 204 | ||
| AY150699.1 | - | 8 | 1376 | |||
| BC118526.1 | - | 8 | 1370 | 40099214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Disp2| Gene Symbol: Disp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170593.3 | NP_733481.1 | 8 | 1575 | ||
| GenBank mRNA | AK122547.1 | - | 1 | 204 | ||
| AY150699.1 | - | 8 | 1376 | |||
| BC118526.1 | - | 8 | 1370 | 40099214 | ||