siRNA Details
| Gene Symbol: Zcchc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153538.3 | NP_705766.3 | 11, 12 | 1710 | ||
| GenBank mRNA | AK031043.1 | - | 10, 11 | 1287 | ||
| BC023880.1 | - | 3, 4 | 421 | 5354757 | ||
| BC043111.1 | - | 11, 12 | 1664 | 6407010 | ||

Zcchc6
Genomic Map