siRNA Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177460.4 | NP_803411.3 | 4, 5 | 801 | ||
| GenBank mRNA | AK033791.1 | - | 751 | |||
| AK133475.1 | - | 4, 5 | 802 | |||
| AK166002.1 | - | 3, 4 | 565 | |||
| BC014731.1 | - | 3, 4 | 831 | |||
| BC055447.1 | - | 4, 5 | 770 | 3594406 | ||

Parp16
Genomic Map