siRNA Details
| Gene Symbol: Tcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130458.1 | NP_001123930.1 | 9 | 1352 | ||
| NM_001130459.1 | NP_001123931.1 | 9 | 1321 | |||
| NM_015749.3 | NP_056564.1 | 9 | 1355 | |||
| GenBank mRNA | AF090686.1 | - | 8 | 1282 | ||
| AK133707.1 | - | 9 | 1317 | |||
| AK143761.1 | - | 9 | 1323 | |||
| AK147176.1 | - | 9 | 1326 | |||
| AK161618.1 | - | 9 | 1322 | |||
| AK170401.1 | - | 9 | 1341 | |||
| BC003720.1 | - | 8 | 1228 | 3601531 | ||

Tcn2
Genomic Map