siRNA Details
Gene Symbol: Tmprss13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013373.2 | NP_001013391.2 | 5 | 680 | ||
GenBank mRNA | BC042878.1 | - | 4 | 416 | ||
BC085323.1 | - | 5 | 665 | 30734093 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmprss13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013373.2 | NP_001013391.2 | 5 | 680 | ||
GenBank mRNA | BC042878.1 | - | 4 | 416 | ||
BC085323.1 | - | 5 | 665 | 30734093 |