siRNA Details
| Gene Symbol: Tmprss13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013373.2 | NP_001013391.2 | 6 | 836 | ||
| GenBank mRNA | BC042878.1 | - | 5 | 572 | ||
| BC085323.1 | - | 6 | 821 | 30734093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss13| Gene Symbol: Tmprss13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013373.2 | NP_001013391.2 | 6 | 836 | ||
| GenBank mRNA | BC042878.1 | - | 5 | 572 | ||
| BC085323.1 | - | 6 | 821 | 30734093 | ||