siRNA Details
Gene Symbol: 4932411E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172534.2 | NP_766122.1 | 2 | 251 | ||
GenBank mRNA | AK029964.1 | - | 2 | 252 | ||
BC140961.1 | - | 2 | 237 | 9055573 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4932411E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172534.2 | NP_766122.1 | 2 | 251 | ||
GenBank mRNA | AK029964.1 | - | 2 | 252 | ||
BC140961.1 | - | 2 | 237 | 9055573 |