siRNA Details
| Gene Symbol: 4930486L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178098.2 | NP_835199.1 | 6 | 755 | ||
| GenBank mRNA | AK145933.1 | - | 6 | 756 | ||
| AY146988.1 | - | 6 | 755 | |||
| BC061218.1 | - | 6 | 742 | 30281034 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930486L24Rik| Gene Symbol: 4930486L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178098.2 | NP_835199.1 | 6 | 755 | ||
| GenBank mRNA | AK145933.1 | - | 6 | 756 | ||
| AY146988.1 | - | 6 | 755 | |||
| BC061218.1 | - | 6 | 742 | 30281034 | ||