siRNA Details
| Gene Symbol: Tcp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085555.1 | NP_001079024.1 | 7 | 953 | ||
| NM_013687.3 | NP_038715.2 | 7 | 1190 | |||
| GenBank mRNA | BC016610.1 | - | 7 | 1173 | ||
| BC058793.1 | - | 6 | 1129 | |||
| BC089527.1 | - | 7 | 1191 | |||
| BC089528.1 | - | 7 | 936 | 6741317 | ||
| X52127.1 | - | 5 | 712 | |||
| X52128.1 | - | 7 | 1168 | |||

Tcp11
Genomic Map