siRNA Details
| Gene Symbol: Iqub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172535.3 | NP_766123.2 | 2 | 261 | ||
| GenBank mRNA | AK029931.1 | - | 2 | 251 | ||
| AK133019.1 | - | 2 | 242 | |||
| BC137937.1 | - | 2 | 228 | 8860956 | ||
| BC137957.1 | - | 2 | 228 | 8860976 | ||

Iqub
Genomic Map