siRNA Details
| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178115.4 | NP_835216.3 | 19 | 2812 | ||
| GenBank mRNA | AK144179.1 | - | 19 | 2771 | ||
| BC024916.1 | - | 7 | 944 | |||
| BC046235.1 | - | 7 | 1162 | 6817045 | ||
| BC072596.1 | - | 19 | 2781 | 6409214 | ||

Edrf1
Genomic Map