siRNA Details
| Gene Symbol: Mmp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152944.1 | NP_694423.1 | 6, 7 | 1423 | ||
| GenBank mRNA | AF507967.1 | - | 6, 7 | 1408 | ||
| AY124569.1 | - | 6, 7 | 1423 | |||
| BC106861.2 | - | 6, 7 | 1423 | 40047279 | ||
| BC111522.1 | - | 5, 6 | 2230 | 40047286 | ||

Mmp21
Genomic Map