siRNA Details
| Gene Symbol: Zfp879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290779.1 | NP_001277708.1 | 3 | 314 | ||
| NM_173387.3 | NP_775563.1 | 4 | 441 | |||
| GenBank mRNA | AK036152.1 | - | 4 | 442 | ||
| BC118537.1 | - | 3 | 308 | 40094138 | ||

Zfp879
Genomic Map