siRNA Details
Gene Symbol: Neurl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153408.2 | NP_700457.1 | 2 | 429 | ||
GenBank mRNA | AF321278.1 | - | 2 | 429 | ||
AK040949.1 | - | 2 | 440 | |||
AK156207.1 | - | 2 | 440 | |||
BC056622.1 | - | 2 | 420 | 4989283 |
