siRNA Details
| Gene Symbol: Wfikkn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100454.1 | NP_001093924.1 | 2 | 1286 | ||
| GenBank mRNA | BC048715.1 | - | 2 | 724 | ||
| BC147298.1 | - | 2 | 504 | 9056349 | ||
| BC147299.1 | - | 2 | 504 | 9056355 | ||

Wfikkn1
Genomic Map